Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzyl alcohol, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
17849-38-6
C7H7ClO
142.58
MFCD00004604
MBYQPPXEXWRMQC-UHFFFAOYSA-N
2-chlorobenzyl alcohol,2-chlorophenyl methanol,benzenemethanol, 2-chloro,2-chlorobenzenemethanol,o-chlorobenzyl alcohol,benzyl alcohol, o-chloro,2-chloro-phenyl-methanol,2-chlorophenyl methan-1-ol,benzenemethanol, ar-chloro,2-chlorobenzylalcohol
28810
(2-chlorophenyl)methanol
C1=CC=C(C(=C1)CO)Cl
17849-38-6
C7H7ClO
142.58
MFCD00004604
MBYQPPXEXWRMQC-UHFFFAOYSA-N
2-chlorobenzyl alcohol,2-chlorophenyl methanol,benzenemethanol, 2-chloro,2-chlorobenzenemethanol,o-chlorobenzyl alcohol,benzyl alcohol, o-chloro,2-chloro-phenyl-methanol,2-chlorophenyl methan-1-ol,benzenemethanol, ar-chloro,2-chlorobenzylalcohol
28810
(2-chlorophenyl)methanol
C1=CC=C(C(=C1)CO)Cl
Color White
Infrared Spectrum Authentic
Boiling Point 227°C
Melting Point 69°C to 72°C
Quantity 50g
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula ClC6H4CH2OH
Beilstein 06, 444
Solubility Information Solubility in water: limited soluble in water. Other solubilities: 1g/10mL in methanol
Formula Weight 142.58
Physical Form Fine Crystalline Powder
Percent Purity 99%
Chemical Name or Material 2-Chlorobenzyl alcohol, 99%
Show More Show Less

EINECSNumber : 241-801-7

TSCA : TSCA