Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrazine hydrate, 55% (Hydrazine, 35%)
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Specifications
| Boiling Point | 109.4°C |
| Melting Point | -65.0°C |
| Physical Form | Liquid |
| Quantity | 100 mL |
| Assay Percent Range | 55% |
| Linear Formula | H2NNH2·xH2O |
| Flash Point | >100°C |
| Fieser | 01,434; 02,211; 03,153; 04,248; 05,327; 06,280; 07,170; 16,174 |
| Merck Index | 14,4771 |
| Solubility Information | Solubility in water: miscible. Other solubilities: miscible with alcohol,insoluble in chloroform and ether |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Fatal if inhaled. May cause cancer. Toxic if swallowed. Toxic in contact with skin. May cause an allergic skin reaction. Very toxic to aquatic life with long lasting effects. Causes severe skin b |
| Precautionary Statement | GHS P Statement Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. Avoid release to the environment. IF IN EYES: Rinse cautiously with water for several m |
| RTECSNumber | MV4590000 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction