Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzyl chloride, 98+%

Catalog No. AC186530010
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle

CAS: 611-19-8 | C7H6Cl2 | 161.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-19-8
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.03
MDL Number MFCD00000893
InChI Key BASMANVIUSSIIM-UHFFFAOYSA-N
Synonym 2-chlorobenzyl chloride, 1-chloro-2-chloromethyl benzene, 2-chlorobenzylchloride, o-chlorobenzyl chloride, alpha,2-dichlorotoluene, benzene, 1-chloro-2-chloromethyl, alpha,o-dichlorotoluene, ar,alpha-dichlorotoluene, ortho-alpha-dichlorotoluene, unii-h58di83rmd
PubChem CID 11906
IUPAC Name 1-chloro-2-(chloromethyl)benzene
SMILES C1=CC=C(C(=C1)CCl)Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -13°C
Color Colorless to Yellow
Density 1.2700g/mL
Boiling Point 213°C to 214°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5580 to 1.56
Linear Formula ClC6H4CH2Cl
Quantity 1 L
Beilstein 05, 297
Specific Gravity 1.27
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in most common solvents (reacts with man
Formula Weight 161.03
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Chlorobenzyl chloride, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Very toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
EINECSNumber 210-258-8
RTECSNumber CZ0195000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.