Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethanol, 99%, Thermo Scientific Chemicals

Catalog No. AA36636K2
Click to view available options
Quantity:
50 mL
250 mL
1 L

2-Chloroethanol is used in the preparation of several dyes by the alkylation of aniline derivatives with chloroethanol. It is a building block in the production of pharmaceuticals and plasticizers. Also used for manufacture of thiodiglycol. It is a solvent for cellulose acetate and ethyl cellulose, textile printing dyes, in dewaxing, refining of rosin, extraction of pine lignin and the cleaning of machines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-07-3
Molecular Formula C2H5ClO
Molecular Weight (g/mol) 80.511
MDL Number MFCD00002829
InChI Key SZIFAVKTNFCBPC-UHFFFAOYSA-N
Synonym ethylene chlorohydrin, ethanol, 2-chloro, glycol chlorohydrin, 2-chloroethyl alcohol, chloroethanol, glycol monochlorohydrin, 2-monochloroethanol, ethylchlorohydrin, 2-chlorethanol, 2-chloro-1-ethanol
PubChem CID 34
ChEBI CHEBI:28200
IUPAC Name 2-chloroethanol
SMILES C(CCl)O

Specifications

Melting Point -63°C
Color Yellow
Density 1.202 g/mL
Boiling Point 129°C
Flash Point 55°C (131°F)
Odor Ether-like
Refractive Index 1.441
Linear Formula ClCH2CH2OH
Quantity 1 L
UN Number UN1135
Beilstein 878139
Merck Index 14,3793
Solubility Information Fully miscible with water.
Formula Weight 80.51
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Chloroethanol
Show More Show Less
Hazard Category H226-H300+H310+H330
Hazard Statement GHS H Statement
H300-H310-H330-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ETHYLENE CHLOROHYDRIN
EINECSNumber 203-459-7
RTECSNumber KK0875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.