Learn More
2-Chloroethylamine hydrochloride, 98+%
Description
2-chloroethylamine hydrochloride is used as an intermediate in active pharmaceutical ingredients and dyes. It is also used in organic synthesis. Further, it is employed as a derivatizing reagent for amino acids, dipeptides and nucleotides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-chloroethylamine hydrochloride is used as an intermediate in active pharmaceutical ingredients and dyes. It is also used in organic synthesis. Further, it is employed as a derivatizing reagent for amino acids, dipeptides and nucleotides.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 143°C to 148°C |
| pH | 2 to 3 |
| Odor | Odorless |
| Quantity | 1000 g |
| UN Number | UN3261 |
| Beilstein | 3590304 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water. |
| Formula Weight | 115.99 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H314-H341 Causes severe skin burns and eye damage. Suspected of causing genetic defects. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| EINECSNumber | 212-793-2 |
| RTECSNumber | KR1800000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.