Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. C0851100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0851100G 100 g
C085125G 25 g
C0851500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0851100G Supplier TCI America Supplier No. C0851100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 870-24-6
Molecular Formula C2H7Cl2N
Molecular Weight (g/mol) 115.99
MDL Number MFCD00012887,MFCD00137399
InChI Key ONRREFWJTRBDRA-UHFFFAOYSA-N
Synonym 2-chloroethylamine hydrochloride, 2-chloroethanamine hydrochloride, 1-amino-2-chloroethane hydrochloride, ethanamine, 2-chloro-, hydrochloride, 2-aminoethyl chloride hydrochloride, ethanamine, 2-chloro-, hydrochloride 1:1, 2-chloroethan-1-amine hydrochloride, chlorethamine hydrochloride, 2-chloro-ethylamine hydrochloride, chlorethamine hcl
PubChem CID 9793737
IUPAC Name 2-chloroethan-1-aminium chloride
SMILES [Cl-].[NH3+]CCCl

Specifications

Melting Point 147°C
Quantity 100 g
Formula Weight 115.99
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloroethylamine Hydrochloride
RTECSNumber KR1800000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.