Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenyl-1,2,3,6-tetrahydropyridine hydrochloride, 99%, Thermo Scientific™

Catalog No. AC158860100
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43064-12-6
Molecular Formula C11H14ClN
Molecular Weight (g/mol) 195.69
MDL Number MFCD00012752
InChI Key POGWXTJNUCZEPR-UHFFFAOYSA-N
Synonym 4-phenyl-1,2,3,6-tetrahydropyridine hydrochloride, 1,2,3,6-tetrahydro-4-phenylpyridine hydrochloride, 4-phenyl-1,2,3,6-tetrahydroxpyridine, pyridine, 1,2,3,6-tetrahydro-4-phenyl-, hydrochloride, 4-phenyl-1,2,5,6-tetrahydropyridine, chloride, acmc-209jsh, 4-phenyl-1,2,3,6-tetra-hydropyridine, 4-phenyl-1,2,3,6-tetrahydropyridine hcl, 4-phenyl-1,2,3,6-tetrahydropyridinehydrochloride, 4-phenyl-1,2,3,6-tetrahydro-pyridine hydrochloride
PubChem CID 2723860
IUPAC Name 4-phenyl-1,2,3,6-tetrahydropyridine;hydrochloride
SMILES C1CNCC=C1C2=CC=CC=C2.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 200°C to 204°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 10 g
Beilstein 20, 3239
Formula Weight 195.69
Percent Purity 98 to 101% Total Base (on dry substance), ≥98.5% (TLC)
Physical Form Powder
Chemical Name or Material 4-Phenyl-1,2,3,6-tetrahydropyridine hydrochloride, 99%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing cancer.
Toxic if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 256-071-5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.