Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Chloromethoxy)ethyltrimethylsilane (stabilized with Diisopropylethylamine) 95.0+%, TCI America™
SDP

Catalog No. C133925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C133925ML 25 mL
C13395ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C133925ML Supplier TCI America Supplier No. C133925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 76513-69-4
Molecular Formula C6H15ClOSi
Molecular Weight (g/mol) 166.72
MDL Number MFCD00009919
InChI Key BPXKZEMBEZGUAH-UHFFFAOYSA-N
Synonym 2-trimethylsilyl ethoxymethyl chloride, 2-chloromethoxy ethyl trimethylsilane, sem-chloride, 2-trimethylsilyl ethoxymethylchloride, sem-cl, 2-chloromethyl 2-trimethylsilyl ethyl ether, semcl, 2-chloromethoxy ethyl-trimethylsilane, 2-trimethysilyl-ethoxymethyl chloride
PubChem CID 2724271
IUPAC Name 2-(chloromethoxy)ethyl-trimethylsilane
SMILES C[Si](C)(C)CCOCCl

Specifications

Color Colorless
Boiling Point 59°C
Quantity 25 mL
UN Number 2920
Formula Weight 166.72
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Chloromethoxy)ethyltrimethylsilane (stabilized with Diisopropylethylamine)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.