Learn More
2-(Chloromethoxy)ethyltrimethylsilane, tech. 90%, stab. with 0.1% N,N-Diisopropylethylamine
Description
2-(Chloromethoxy)ethyltrimethylsilane is used in the preparation of SEM [2-(trimethylsilyl)ethoxy]methyl]-ethers. It serves as a phenol protecting group in the synthesis of laterifluorones. Further, it reacts with 1H-imidazole to prepare 1-(2-trimethylsilanyl-ethoxymethyl)-1H-imidazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-(Chloromethoxy)ethyltrimethylsilane is used in the preparation of SEM [2-(trimethylsilyl)ethoxy]methyl]-ethers. It serves as a phenol protecting group in the synthesis of laterifluorones. Further, it reacts with 1H-imidazole to prepare 1-(2-trimethylsilanyl-ethoxymethyl)-1H-imidazole.
Notes
Store in a cool place. Moisture sensitive. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Boiling Point | 170°C to 172°C |
| Quantity | 25 g |
| Linear Formula | (CH3)3SiCH2CH2OCH2Cl |
| UN Number | UN2920 |
| Beilstein | 3587289 |
| Flash Point | 46°C (115°F) |
| Formula Weight | 166.72 |
| Percent Purity | 90% |
| Grade | Technical |
| Refractive Index | 1.435 |
| Show More |
Safety and Handling
| Hazard Category | H226-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H226 Causes severe skin burns and eye damage. Causes serious eye damage. Flammable liquid and vapor. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S. |
| EINECSNumber | 278-483-4 |
| TSCA | No |
| Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.