Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Chloromethoxy)ethyltrimethylsilane, tech. 90%, stab. with 0.1% N,N-Diisopropylethylamine

Catalog No. p-7036733
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 76513-69-4 | C6H15ClOSi | 166.72 g/mol

2-(Chloromethoxy)ethyltrimethylsilane is used in the preparation of SEM [2-(trimethylsilyl)ethoxy]methyl]-ethers. It serves as a phenol protecting group in the synthesis of laterifluorones. Further, it reacts with 1H-imidazole to prepare 1-(2-trimethylsilanyl-ethoxymethyl)-1H-imidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Chloromethoxy)ethyltrimethylsilane is used in the preparation of SEM [2-(trimethylsilyl)ethoxy]methyl]-ethers. It serves as a phenol protecting group in the synthesis of laterifluorones. Further, it reacts with 1H-imidazole to prepare 1-(2-trimethylsilanyl-ethoxymethyl)-1H-imidazole.

Notes
Store in a cool place. Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76513-69-4
Molecular Formula C6H15ClOSi
Molecular Weight (g/mol) 166.72
MDL Number MFCD00009919
InChI Key BPXKZEMBEZGUAH-UHFFFAOYSA-N
Synonym 2-trimethylsilyl ethoxymethyl chloride, 2-chloromethoxy ethyl trimethylsilane, sem-chloride, 2-trimethylsilyl ethoxymethylchloride, sem-cl, 2-chloromethyl 2-trimethylsilyl ethyl ether, semcl, 2-chloromethoxy ethyl-trimethylsilane, 2-trimethysilyl-ethoxymethyl chloride
PubChem CID 2724271
IUPAC Name 2-(chloromethoxy)ethyl-trimethylsilane
SMILES C[Si](C)(C)CCOCCl

Specifications

Boiling Point 170°C to 172°C
Quantity 1 g
Linear Formula (CH3)3SiCH2CH2OCH2Cl
UN Number UN2920
Beilstein 3587289
Flash Point 46°C (115°F)
Formula Weight 166.72
Percent Purity 90%
Grade Technical
Refractive Index 1.435
Sensitivity Moisture sensitive
Density 0.942
Chemical Name or Material 2-(Chloromethoxy)ethyltrimethylsilane, stabilized with 0.3% N-ethyldiisopropylamine
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 278-483-4
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.