Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Chloromethyl)-1-methyl-1H-imidazole hydrochloride, 97%, Thermo Scientific™

Catalog No. CC20874CB
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 78667-04-6
Molecular Formula C5H8Cl2N2
Molecular Weight (g/mol) 167.033
InChI Key JDIPKQBKSBWIKU-UHFFFAOYSA-N
Synonym 2-chloromethyl-1-methyl-1h-imidazole hydrochloride, 2-chloromethyl-1-methylimidazole hydrochloride, 1h-imidazole, 2-chloromethyl-1-methyl-, monohydrochloride, 2-chloromethyl-1-methyl-1h-imidazole hcl, 1-methyl-2-chloromethylimidazole hydrochloride, 2-chloromethyl-1-methylimidazole, chloride, 2-chloromethyl-1-methyl imidazole hydrochloride, 1-methyl-2-chloromethyl imidazole hydrochloride
PubChem CID 2795406
IUPAC Name 2-(chloromethyl)-1-methylimidazole;hydrochloride
SMILES CN1C=CN=C1CCl.Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 167.04
Percent Purity 97%
Chemical Name or Material 2-(Chloromethyl)-1-methyl-1H-imidazole hydrochloride
Hazard Category GHS Signal Word: Danger
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.