Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Chloromethyl)-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. C253225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C253225G 25 g
C25325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C253225G Supplier TCI America Supplier No. C253225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 127337-60-4
Molecular Formula C9H10Cl2F3NO
Molecular Weight (g/mol) 276.08
MDL Number MFCD00800224
InChI Key CMZBQUWICURDCD-UHFFFAOYSA-N
Synonym 2-chloromethyl-3-methyl-4-2,2,2-trifluoroethoxy pyridine hydrochloride, 2-chloromethyl-3-methyl-4-2,2,2-trifluoro-ethoxy-pyridine hcl, 2-chloromethyl-3-methyl-4-2,2,2-trifluoroethoxy pyridine, hydrochloride, 2-chloromethyl-3-methyl-4-3-meyhoxylpropanoxyl pyridine hydrochloride, 2-chloromethyl-3-methyl-4-2,2,2-trifluoroethoxy pyridine;hydrochloride, 2-chloromethyl-3-methyl-4-2,2,2-trifluoro-ethoxy-pyridine hydrochloride, 1-2-chloromethyl-3-methyl 4-pyridyloxy-2,2,2-trifluoroethane, chloride, pubchem14161, acmc-209bce
PubChem CID 16217647
IUPAC Name 2-(chloromethyl)-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine hydrochloride
SMILES Cl.CC1=C(OCC(F)(F)F)C=CN=C1CCl

Specifications

Melting Point 214°C
Color White-Yellow
Quantity 25 g
Formula Weight 276.08
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Chloromethyl)-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine Hydrochloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.