Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Chloromethyl)benzimidazole, 95%

Catalog No. AAB2554806
Click to view available options
Quantity:
5 g

CAS: 4857-04-9 | C8H7ClN2 | 166.61 g/mol

2-(Chloromethyl)benzimidazole is used as intermediate in pharmaceutical , in organic synthesis .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Chloromethyl)benzimidazole is used as intermediate in pharmaceutical , in organic synthesis .

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4857-04-9

Specifications

Melting Point 141°C to 143°C (decomposition)
Molecular Formula C8H7ClN2
MDL Number MFCD00005602
Quantity 5 g
Synonym 2-chloromethylbenzimidazole, 2-chloromethyl benzimidazole, 2-chloromethyl-1h-benzo d imidazole, 2-chloromethyl-1h-benzimidazole, 2-chloromethyl-1h-1,3-benzodiazole, 2-chloromethyl-1h-benzoimidazole, 1h-benzimidazole, 2-chloromethyl, dimezole-7, 2-chloromethyl-1h-1,3-benzimidazole
Solubility Information Insoluble in water.
InChI Key SPMLMLQATWNZEE-UHFFFAOYSA-N
SMILES ClCC1=NC2=CC=CC=C2N1
IUPAC Name 2-(chloromethyl)-1H-benzimidazole
Molecular Weight (g/mol) 166.61
PubChem CID 78571
Formula Weight 166.61
Percent Purity 96%
Chemical Name or Material 2-(Chloromethyl)benzimidazole
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 225-452-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.