Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloronaphthalene, 99%

Catalog No. p-89634
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 91-58-7 | C10H7Cl | 162.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-58-7
Molecular Formula C10H7Cl
Molecular Weight (g/mol) 162.62
MDL Number MFCD00035731
InChI Key CGYGETOMCSJHJU-UHFFFAOYSA-N
Synonym naphthalene, 2-chloro, 2-chloro naphthalene, beta-chloronaphthalene, halowax, 2-chlornaftalen, rcra waste number u047, 2-chloro-naphthalene, .beta.-chloronaphthalene, 2-chlornaftalen czech, unii-49o81u3iti
PubChem CID 7056
IUPAC Name 2-chloronaphthalene
SMILES C1=CC=C2C=C(C=CC2=C1)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 57°C to 60°C
Color White
Boiling Point 256°C
Flash Point 125°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 2152
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in ethanol,benzene,ether and chloroform
Formula Weight 162.62
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloronaphthalene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-079-9
RTECSNumber QJ2275000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.