Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chlorophenol

Catalog No. C62794100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Change view
Click to view available options
Quantity:
100 g
1 kg
2 kg
Catalog No. Quantity
C62794100G 100 g
C627941KG 1 kg
C627942KG 2 kg
3 options

Catalog No. C62794100G

Supplier: Sigma Aldrich C62794100G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Chlorophenol

Specifications

CAS 95-57-8
Melting Point 8°C (lit.)
Density 1.241 g/mL (at 25°C (literature))
Boiling Point 175°C to 176°C (lit.)
Molecular Formula C6H5ClO
Refractive Index n20/D 1.558 (literature)
Linear Formula ClC6H4OH
MDL Number MFCD00002159
Quantity 100 g
Molecular Weight (g/mol) 128.56
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.