Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorophenyl phosphorodichloridate, 98+%

Catalog No. AAA1150522
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 15074-54-1 | C6H4Cl3O2P | 245.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15074-54-1
Molecular Formula C6H4Cl3O2P
Molecular Weight (g/mol) 245.42
MDL Number MFCD00009704
InChI Key VLDPXPPHXDGHEW-UHFFFAOYSA-N
Synonym 2-chlorophenyl phosphorodichloridate, 2-chlorophenyl dichlorophosphate, o-chlorophenyl dichlorophosphate, phosphorodichloridic acid, 2-chlorophenyl ester, o-chlorophenyl phosphorodichloridate, 2-chlorophenyl phosphoric acid dichloride, 2-chlorophenyl chlorophosphonochloridate, acmc-20anxn, o-chlorophenylphosphonyl dichloride, 2-chlorophenyl dichloridophosphate #
PubChem CID 84775
IUPAC Name 1-chloro-2-dichlorophosphoryloxybenzene
SMILES C1=CC=C(C(=C1)OP(=O)(Cl)Cl)Cl

Specifications

Density 1.491
Boiling Point 82°C to 84°C (0.4 mmHg)
Flash Point 105°C (221°F)
Refractive Index 1.538
Quantity 100 g
UN Number UN3265
Beilstein 911009
Sensitivity Moisture sensitive
Formula Weight 245.43
Percent Purity ≥98%
Chemical Name or Material 2-Chlorophenyl phosphorodichloridate
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 239-129-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.