Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorophenylacetone, 96%

Catalog No. p-7045212
Click to view available options
Quantity:
1 g
5 g

CAS: 6305-95-9 | C9H9ClO | 168.62 g/mol

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Difficult to mix in water.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6305-95-9
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00045175
InChI Key LWGNDIMNCPMZOF-UHFFFAOYSA-N
Synonym 2-chlorophenylacetone, 1-2-chlorophenyl propan-2-one, 1-2-chlorophenyl acetone, 2-propanone, 1-2-chlorophenyl, acmc-20apcv, 2-chlorphenyl acetone, 1-2-chlorophenyl acetone #, 1-2-chlorophenyl-2-propanone, 1-2-chloro-phenyl-propan-2-one
PubChem CID 229355
IUPAC Name 1-(2-chlorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=CC=C1Cl

Specifications

Boiling Point 102°C to 103°C (3 mmHg)
Refractive Index 1.536
Quantity 1 g
Beilstein 2043542
Solubility Information Difficult to mix in water.
Formula Weight 168.62
Percent Purity 96%
Chemical Name or Material 2-Chlorophenylacetone
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.