Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloropyridine-3-boronic acid, 97%, Thermo Scientific™

Catalog No. AC430640050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 381248-04-0
Molecular Formula C5H5BClNO2
Molecular Weight (g/mol) 157.36
MDL Number MFCD03094997
InChI Key VRDAOVQZVXYRNH-UHFFFAOYSA-N
Synonym 2-chloropyridine-3-boronic acid, 2-chloropyridin-3-yl boronic acid, 2-chloro-3-pyridylboronic acid, 2-chloro-3-pyridineboronic acid, 2-chloropyridine-3-boronicacid, 2-chloropyridin-3-yl-3-boronic acid, 2-chloro-3-pyridyl boronic acid, boronic acid, 2-chloro-3-pyridinyl, pubchem5078
PubChem CID 2762704
IUPAC Name (2-chloropyridin-3-yl)boronic acid
SMILES OB(O)C1=CC=CN=C1Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 116.0°C to 120.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 157.37
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloropyridine-3-boronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.