Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Chloropyridine 98.0+%, TCI America™

Click to view available options
Quantity:
25 g
100 g
500 g
Specifications
Specifications
Color | Yellow |
pH | 6 to 7 (27g/L H2O 20°C) |
Boiling Point | 170°C |
Quantity | 100 g |
UN Number | 2822 |
Formula Weight | 113.54 |
Percent Purity | ≥98.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | 2-Chloropyridine |
Safety and Handling
EINECSNumber | (5)-0720 |
RTECSNumber | US5950000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction