Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorostyrene (stabilized with TBC) 96.0+%, TCI America™
SDP

Catalog No. C028825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C028825G 25 g
C02885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C028825G Supplier TCI America Supplier No. C028825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2039-87-4
Molecular Formula C8H7Cl
Molecular Weight (g/mol) 138.594
MDL Number MFCD00000567
InChI Key ISRGONDNXBCDBM-UHFFFAOYSA-N
Synonym 2-chlorostyrene, 1-chloro-2-vinylbenzene, o-chlorostyrene, benzene, 1-chloro-2-ethenyl, styrene, o-chloro, ortho-chlorostyrene, stryene, o-chloro, styrene, 2-chloro, unii-za43r4q315, ccris 4598
PubChem CID 14906
IUPAC Name 1-chloro-2-ethenylbenzene
SMILES C=CC1=CC=CC=C1Cl

Specifications

Melting Point -63°C
Color Yellow
Boiling Point 187°C
Quantity 25 g
UN Number 1993
Formula Weight 138.59
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorostyrene (stabilized with TBC)
EINECSNumber (3)-3441
RTECSNumber WL4160000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.