Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chlorothiobenzamide, 98%, Thermo Scientific™

Catalog No. AC374410010
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15717-17-6
Molecular Formula C7H6ClNS
Molecular Weight (g/mol) 171.642
InChI Key FLQYOORLPNYQEV-UHFFFAOYSA-N
Synonym 2-chlorothiobenzamide, 2-chlorobenzothioamide, benzenecarbothioamide, 2-chloro, 2-chlorobenzene-1-carbothioamide, o-chlorothiobenzamide, 2-chloro-thiobenzamide, acmc-1btqa, benzamide, o-chlorothio, 2-chloranylbenzenecarbothioamide, benzenecarbothioamide,2-chloro
PubChem CID 2734824
IUPAC Name 2-chlorobenzenecarbothioamide
SMILES C1=CC=C(C(=C1)C(=S)N)Cl

Specifications

Melting Point 62°C to 65°C
Color Yellow
CAS Min % 97.5
CAS Max % 100.0
Quantity 1 g
Infrared Spectrum Authentic
Physical Form Powder
Assay Percent Range 98%
Linear Formula ClC6H4CSNH2
Formula Weight 171.65
Percent Purity ≥97.5%
Chemical Name or Material 2-Chlorothiobenzamide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.