Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyano-N,N-dimethylacetamide 98.0+%, TCI America™
SDP

Catalog No. p-7125173
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 7391-40-4
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.13
MDL Number MFCD00051623
InChI Key MATJPVGBSAQWAC-UHFFFAOYSA-N
Synonym n,n-dimethylcyanoacetamide, acetamide, 2-cyano-n,n-dimethyl, n,n-dimethyl cyanoacetamide, n,n-dimethyl-2-cyanoacetamide, acmc-209osw, n,n-dimethylcyanoacetate, 2-cyano-n,n-dimethyl-acetamide, 2-cyano-n,n-dimethyl-ethanamide, acetamide,2-cyano-n,n-dimethyl, cyanoacetic acid n,n-dimethylamide
PubChem CID 251143
IUPAC Name 2-cyano-N,N-dimethylacetamide
SMILES CN(C)C(=O)CC#N

Specifications

Melting Point 66°C
Quantity 5 g
Formula Weight 112.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyano-N,N-dimethylacetamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.