Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyanoacetamide, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
107-91-5
C3H4N2O
84.08
MFCD00008024
DGJMPUGMZIKDRO-UHFFFAOYSA-N
cyanoacetamide,acetamide, 2-cyano,cyanacetamide,malonamide nitrile,malonamonitrile,nitrilomalonamide,cyanoiminoacetic acid,3-nitrilo-propionamide,propionamide, 3-nitrilo,usaf kf-14
7898
2-cyanoacetamide
C(C#N)C(=O)N
107-91-5
C3H4N2O
84.08
MFCD00008024
DGJMPUGMZIKDRO-UHFFFAOYSA-N
cyanoacetamide,acetamide, 2-cyano,cyanacetamide,malonamide nitrile,malonamonitrile,nitrilomalonamide,cyanoiminoacetic acid,3-nitrilo-propionamide,propionamide, 3-nitrilo,usaf kf-14
7898
2-cyanoacetamide
C(C#N)C(=O)N
Infrared Spectrum Authentic
Flash Point 215°C
Packaging Plastic bottle
Color White to Cream
Melting Point 118°C to 122°C
Quantity 500g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula NCCH2CONH2
Beilstein 02, 589
Merck Index 15, 2678
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 84.08
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 2-Cyanoacetamide, 99%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove

GHS Signal Word: Warning

EINECSNumber : 203-531-8

RTECSNumber : AB5950000

TSCA : TSCA