Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyanobenzoic acid, 94%

Catalog No. p-7033183
Click to view available options
Quantity:
1 g
5 g

Catalog No. AAH5023703

Supplier: Thermo Scientific Chemicals H5023703

Only null left
Add to Cart

CAS: 3839-22-3 | C8H5NO2 | 147.133 g/mol

2-Cyanobenzoic acid has been used as reactant in the synthesis of T-box riboswitch antiterminator RNA-binding oxazolidinone derivatives, Benzoate modified β-cyclodextrin derivatives and Pyrrolo[1,2-f]triazines for use as JAK2 inhibitors. As a reactant involved in decarboxylation, decarboxylative halogenation and protodecarboxylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyanobenzoic acid has been used as reactant in the synthesis of T-box riboswitch antiterminator RNA-binding oxazolidinone derivatives, Benzoate modified β-cyclodextrin derivatives and Pyrrolo[1,2-f]triazines for use as JAK2 inhibitors. As a reactant involved in decarboxylation, decarboxylative halogenation and protodecarboxylation.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3839-22-3
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.133
MDL Number MFCD00045796
InChI Key DTNSDCJFTHMDAK-UHFFFAOYSA-N
Synonym o-cyanobenzoic acid, cyanobenzoic acid, benzoic acid, 2-cyano, 2-carboxybenzonitrile, 2-carboxy-benzonitrile, 2-cyanobenzoicacid, 2-cyano-benzoic acid, pubchem12513, acmc-1bn2v
PubChem CID 138061
IUPAC Name 2-cyanobenzoic acid
SMILES C1=CC=C(C(=C1)C#N)C(=O)O

Specifications

Melting Point 210°C to 212°C
Quantity 1 g
Formula Weight 147.13
Percent Purity 96%
Chemical Name or Material 2-Cyanobenzoic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.