Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyclohexylcyclohexanone 97.0+%, TCI America™
SDP

Catalog No. C061225G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 90-42-6
Molecular Formula C12H20O
Molecular Weight (g/mol) 180.29
MDL Number MFCD00013795
InChI Key UOBQDYFTAJKQAL-UHFFFAOYNA-N
PubChem CID 7016
IUPAC Name [1,1'-bi(cyclohexane)]-2-one
SMILES O=C1CCCCC1C1CCCCC1

Specifications

Color Yellow
Boiling Point 269°C
Quantity 25 g
Formula Weight 180.29
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyclohexylcyclohexanone
RTECSNumber DT7400000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.