Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyclopropyl-L-glycine, 97%

Catalog No. AAH6575506
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 49606-99-7 | C5H9NO2 | 115.13 g/mol

2-Cyclopropyl-L-glycine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyclopropyl-L-glycine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Store at 4°C. Incompatible with heat and oxidizing agents. Protect from heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49606-99-7
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD06659116
InChI Key BUSBCPMSNBMUMT-BYPYZUCNSA-N
Synonym l-cyclopropylglycine, s-2-amino-2-cyclopropylacetic acid, h-cyclopropyl-gly-oh, 2-cyclopropyl-l-glycine, l-cyclopropyl glycine, 2s-2-amino-2-cyclopropylacetic acid, s-amino-cyclopropyl-acetic acid, alpha-cyclopropylglycine, s-amino cyclopropyl acetic acid
PubChem CID 1501944
SMILES [NH3+][C@@H](C1CC1)C([O-])=O

Specifications

Color White
Beilstein 5728866
Solubility Information Soluble in water. Insoluble in ethanol.
IUPAC Name (2S)-2-amino-2-cyclopropylacetic acid
Quantity 5 g
Formula Weight 115.13
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material 2-Cyclopropyl-L-glycine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.