Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Decanone, 97%

Catalog No. AAA1414514
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 693-54-9 | C10H20O | 156.269 g/mol

2-Decanone was used in the synthesis of brushed block copolymers via conjugation through an acid-labile hydrazone linker to poly(ethylene glycol)-poly(aspartate hydrazide) block copolymers; methyl ketones. 2-decanone used as a solvent and an intermediate in the synthesis of pharmaceuticals and agrochemicals. It is also used in perfumes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Decanone was used in the synthesis of brushed block copolymers via conjugation through an acid-labile hydrazone linker to poly(ethylene glycol)-poly(aspartate hydrazide) block copolymers; methyl ketones. 2-decanone used as a solvent and an intermediate in the synthesis of pharmaceuticals and agrochemicals. It is also used in perfumes.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-54-9
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00009571
InChI Key ZAJNGDIORYACQU-UHFFFAOYSA-N
Synonym 2-decanone, methyl octyl ketone, methyl n-octyl ketone, octyl methyl ketone, unii-gx543olt0r, n-c8h17coch3, gx543olt0r, chembl47127
PubChem CID 12741
ChEBI CHEBI:77929
IUPAC Name decan-2-one
SMILES CCCCCCCCC(=O)C

Specifications

Melting Point 3°C
Density 0.824
Boiling Point 210°C to 212°C
Flash Point 86°C (187°F)
Refractive Index 1.425
Quantity 25 g
Beilstein 1747463
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 156.27
Percent Purity 97%
Chemical Name or Material 2-Decanone
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 211-752-6
RTECSNumber HE0725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.