Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Decylthiophene 97.0+%, TCI America™
SDP

Catalog No. D52401ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52401ML 1 mL
D52405ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52401ML Supplier TCI America Supplier No. D52401ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24769-39-9
Molecular Formula C14H24S
Molecular Weight (g/mol) 224.406
MDL Number MFCD00715761
InChI Key GOTHKCARNFTUSW-UHFFFAOYSA-N
PubChem CID 566331
IUPAC Name 2-decylthiophene
SMILES CCCCCCCCCCC1=CC=CS1

Specifications

Color Yellow
Boiling Point 167°C
Quantity 1 mL
Formula Weight 224.41
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Decylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.