Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals 2'-Deoxyguanosine 5'-monophosphate, sodium salt hydrate, 99%

Catalog No. AC226340010
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226340010 1 g Glass bottle
AC226342500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC226340010 Supplier Thermo Scientific Chemicals Supplier No. 226340010
Add to Cart

CAS: 52558-16-4 | C10H12N5Na2O7P | 391.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52558-16-4
Molecular Formula C10H12N5Na2O7P
Molecular Weight (g/mol) 391.19
MDL Number MFCD00150780
InChI Key CTPAMSRBXKGZCJ-WJQTVUBHNA-L
Synonym deoxyguanosine monophosphate tetrahydrate sodium salt
PubChem CID 131854716
IUPAC Name [(2R,3S,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium;tetrahydrate
SMILES [Na+].[Na+].NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](COP([O-])([O-])=O)O2)C(=O)N1

Specifications

Absorbance 0.65 to 0.68 (A280/A260) c=1,water,0.25 to 0.28 (A290/A260) c=1,water
CAS Min % 98.5
CAS Max % 100.0
Melting Point >245°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 391.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2′-Deoxyguanosine 5′-monophosphate,sodium salt hydrate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.