Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Diethylaminoethanol, Reagent, 98%, Spectrum™ Chemical
SDP

Catalog No. 18600109
Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can

D1016, 100-37-8, C6H15NO

Spectrum™ Chemical 2-Diethylaminoethanol, Reagent is used as a precursor chemical to procaine. In condensate and steam lines, it is used as a corrosion inhibitor because it neutralizes carbonic acid and scavenging oxygen. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 100-37-8
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.19
InChI Key BFSVOASYOCHEOV-UHFFFAOYSA-N
IUPAC Name 2-(diethylamino)ethan-1-ol
SMILES CCN(CC)CCO

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 500 mL
UN Number UN2686
Percent Purity 98%
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.