Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxybenzamide, 97%

Catalog No. AAB2545536
Click to view available options
Quantity:
100 g
500 g

CAS: 938-73-8 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938-73-8
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00007977
InChI Key SBNKFTQSBPKMBZ-UHFFFAOYSA-N
Synonym ethenzamide, benzamide, 2-ethoxy, o-ethoxybenzamide, ethbenzamide, etenzamide, ethenzamid, etamide, ethosalicyl, etosalicil, etosalicyl
PubChem CID 3282
IUPAC Name 2-ethoxybenzamide
SMILES CCOC1=CC=CC=C1C(N)=O

Specifications

Melting Point 131°C to 134°C
Quantity 500 g
Merck Index 14,3731
Formula Weight 165.19
Percent Purity 97%
Chemical Name or Material 2-Ethoxybenzamide
EINECSNumber 213-346-4
RTECSNumber CU4381000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.