Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxyethyl acetate, 98+%, Thermo Scientific Chemicals

Catalog No. AAA136390F
Change view
Click to view available options
Quantity:
500 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA136390F 2500 mL
AAA13639AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA136390F Supplier Thermo Scientific Chemicals Supplier No. A136390F
Add to Cart

2-Ethoxyethyl acetate is used as a solvent dissolve resin, leather, ink, for the formulation of paints, lacquers and varnishes for industrial use. It is also used to study the field evaluation of a passive badge for measuring 2-ethoxyethyl acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxyethyl acetate is used as a solvent dissolve resin, leather, ink, for the formulation of paints, lacquers and varnishes for industrial use. It is also used to study the field evaluation of a passive badge for measuring 2-ethoxyethyl acetate.

Solubility
Soluble in water(230g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-15-9
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00009251
InChI Key SVONRAPFKPVNKG-UHFFFAOYSA-N
Synonym ethylene glycol monoethyl ether acetate, cellosolve acetate, oxitol acetate, ethoxyethyl acetate, oxytol acetate, ethylglycol acetate, ethyl cellosolve acetate, 1-acetoxy-2-ethoxyethane, ethylglykolacetat, octan etoksyetylu
PubChem CID 8095
IUPAC Name 2-ethoxyethyl acetate
SMILES CCOCCOC(C)=O

Specifications

Melting Point -62°C
Density 0.975
Boiling Point 155°C to 157°C
Flash Point 52°C (126°F)
Odor Fruit-like
Refractive Index 1.407
Linear Formula CH3CO2CH2CH2OCH2CH3
Quantity 2500 mL
UN Number UN1172
Beilstein 1748677
Merck Index 14,3751
Solubility Information Soluble in water(230g/L).
Formula Weight 132.16
Percent Purity ≥98%
Chemical Name or Material 2-Ethoxyethyl acetate
Show More Show Less
Hazard Category H226-H302+H312+H332-H360FD
Hazard Statement GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P308+P313-P312-P330-P363-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOETHYL ETHER ACETATE
EINECSNumber 203-839-2
RTECSNumber KK8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.