Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxyethyl Benzoate 98.0+%, TCI America™
SDP

Catalog No. B09555ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09555ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B09555ML Supplier TCI America Supplier No. B09555ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5451-72-9
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00059371
InChI Key KPHLTQOKDPSIGL-UHFFFAOYSA-N
Synonym Benzoic Acid 2-Ethoxyethyl Ester, Ethylene Glycol Monoethyl Ether Benzoate, Cellosolve Benzoate
PubChem CID 228742
IUPAC Name 2-ethoxyethyl benzoate
SMILES CCOCCOC(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 261°C
Quantity 5 mL
Formula Weight 194.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethoxyethyl Benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.