Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxyethyl Isobutyrate 98.0+%, TCI America™
SDP

Catalog No. I028225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I028225ML 25 mL
I02825ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I028225ML Supplier TCI America Supplier No. I028225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54396-97-3
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.21
MDL Number MFCD00009250
InChI Key ANSOCBLHUNAGRK-UHFFFAOYSA-N
Synonym Cellosolve Isobutyrate, Isobutyric Acid 2-Ethoxyethyl Ester, Ethylene Glycol Monoethyl Ether Isobutyrate
PubChem CID 108587
IUPAC Name 2-ethoxyethyl 2-methylpropanoate
SMILES CCOCCOC(=O)C(C)C

Specifications

Color Yellow
Boiling Point 85°C
Quantity 25 mL
UN Number 3272
Formula Weight 160.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethoxyethyl Isobutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.