Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Ethoxyphenol 99.0+%, TCI America™

Click to view available options
Quantity:
25 g
500 g
Specifications
Specifications
Color | White |
Boiling Point | 216°C |
Quantity | 25 g |
Formula Weight | 138.17 |
Percent Purity | ≥99.0% (GC) |
Physical Form | Crystalline Lumps |
Chemical Name or Material | 2-Ethoxyphenol |
Safety and Handling
EINECSNumber | (3)-0568 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction