Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-1,3-cyclopentanedione 97.0+%, TCI America™
SDP

Catalog No. E050725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E050725G 25 g
E05075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E050725G Supplier TCI America Supplier No. E050725G

Chemical Identifiers

CAS 823-36-9
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00044191
InChI Key YDFBIBUYOUFJMR-UHFFFAOYSA-N
Synonym 2-ethyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-ethyl, unii-91a82te3os, acmc-209poa, 2-ethyl-cyclopentan-1,3-dione, 2-ethyl-cyclopentane-1,3-dione
PubChem CID 69978
IUPAC Name 2-ethylcyclopentane-1,3-dione
SMILES CCC1C(=O)CCC1=O

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 25 g
Formula Weight 126.16
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethyl-1,3-cyclopentanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.