Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-1-butanol, 99%

Catalog No. p-7025514
Click to view available options
Quantity:
500 mL
2500 mL

CAS: 97-95-0 | C6H14O | 102.18 g/mol

It is used as a perfuming agent in cosmetics industry. 2-Ethyl-1-butanol is used for the synthesis of various pharmaceutical compounds, such as novel branched Alkyl carbamates, acting as anticonvulsant agent. It is also a solvent used for various organic synthesis, separation processes and ionic liquids, and solvent mixtures for pharmaceutical applications . It is also used to make penetrating oils, corrosion inhibitors, plasticizers, and perfumes; as cleaning agent for printed circuits; and to improve flow of paints and varnishes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a perfuming agent in cosmetics industry. 2-Ethyl-1-butanol is used for the synthesis of various pharmaceutical compounds, such as novel branched Alkyl carbamates, acting as anticonvulsant agent. It is also a solvent used for various organic synthesis, separation processes and ionic liquids, and solvent mixtures for pharmaceutical applications . It is also used to make penetrating oils, corrosion inhibitors, plasticizers, and perfumes; as cleaning agent for printed circuits; and to improve flow of paints and varnishes.

Solubility
Soluble in water (4 mg/ml at 25°C), alcohol, ether, and most organic solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-95-0
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00004744
InChI Key TZYRSLHNPKPEFV-UHFFFAOYSA-N
Synonym 2-ethyl-1-butanol, 1-butanol, 2-ethyl, 2-ethylbutanol, 2-ethylbutyl alcohol, pseudohexyl alcohol, 3-methylolpentane, 2-ethylbutanol-1, ethylbutanol, 3-pentylcarbinol, sec-pentylcarbinol
PubChem CID 7358
IUPAC Name 2-ethylbutan-1-ol
SMILES CCC(CC)CO

Specifications

Melting Point -52°C
Density 0.831
Boiling Point 146°C to 148°C
Flash Point 58°C (136°F)
Refractive Index 1.422
Quantity 500 mL
UN Number UN1987
Beilstein 1731254
Solubility Information Soluble in water (4mg/ml at 25°C),alcohol,ether,and most organic solvents.
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material 2-Ethyl-1-butanol
Show More Show Less
Hazard Category H226-H302+H312
Hazard Statement GHS H Statement
H311-H226-H302
Toxic in contact with skin.
Flammable liquid and vapour.
Harmful if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 202-621-4
RTECSNumber EL3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.