Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-1-butene, 97%

Catalog No. p-7045672
Click to view available options
Quantity:
5 g
25 g

CAS: 760-21-4 | C6H12 | 84.162 g/mol

2-Ethyl-1-butene is used as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyl-1-butene is used as an intermediate in organic synthesis and pharmaceuticals.

Solubility
Soluble in alcohol, and methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 760-21-4
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009341
InChI Key RYKZRKKEYSRDNF-UHFFFAOYSA-N
Synonym 2-ethyl-1-butene, 3-methylenepentane, pentane, 3-methylene, 2-ethylbut-1-ene, 1-butene, 2-ethyl, unii-o5mh5v756e, 1,1-diethylethene, acmc-1blm9, 1-butene, 2-ethyl-8ci
PubChem CID 12970
IUPAC Name 3-methylidenepentane
SMILES CCC(=C)CC

Specifications

Melting Point -132°C
Density 0.689
Boiling Point 63°C to 64°C
Flash Point −26°C (−14°F)
Refractive Index 1.396
Quantity 5 g
UN Number UN3295
Beilstein 1697102
Solubility Information Soluble in alcohol,and methanol. Insoluble in water.
Formula Weight 84.16
Percent Purity 97%
Chemical Name or Material 2-Ethyl-1-butene
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 212-078-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.