Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-1-hexanol, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187393 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
5 mL

2-Ethyl-1-hexanol is an industrial chemical widely used in the production of polyvinyl chloride(PVC) plasticizers and is a potent indoor air pollutant emitted mainly from furniture or carpets with a PVC backing.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 104-76-7
Ignition Point 550°F
Vapor Pressure 0.2 mmHg (20°C)
Vapor Density 4.49 (vs. air)
Density 0.833 g/mL (at 25°C)
Flash Point 75°C - closed cup
Linear Formula CH3(CH2)3 CH(C2H5)CH2 OH
MDL Number MFCD00004746
Melting Point -76°C
Boiling Point 183°C to 186°C
Synonym Isooctyl alcohol
Type Analytical Standard
Quantity 1 mL
Formula Weight 130.23
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.