Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-1-hexanol, 99%

Catalog No. AC118530025
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum

CAS: 104-76-7 | C8H18O | 130.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-76-7
MDL Number MFCD00004746
InChI Key YIWUKEYIRIRTPP-UHFFFAOYSA-N
Synonym 2-ethylhexanol, 2-ethyl-1-hexanol, 1-hexanol, 2-ethyl, 2-ethylhexyl alcohol, ethylhexanol, alcohol, 2-ethylhexyl, xi-2-ethyl-1-hexanol, 2-aethylhexanol german, ethylhexanol, 2, fema no. 3151
PubChem CID 7720
ChEBI CHEBI:16011
IUPAC Name 2-ethylhexan-1-ol
SMILES CCCCC(CC)CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -76.0°C
Density 0.8330g/mL
Boiling Point 184.0°C
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C8H18O
Refractive Index 1.4300 to 1.4330
Linear Formula CH3(CH2)3CH(C2H5)CH2OH
Quantity 2.5 L
Specific Gravity 0.833
Merck Index 15, 3863
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: soluble in many organic solvents including, ethyl alcohol and ethyl ether
Molecular Weight (g/mol) 130.23
Viscosity 10 mPa.s (20°C)
Formula Weight 130.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Ethyl-1-hexanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 203-234-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.