Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Ethylamino)ethanol 98.0+%, TCI America™
SDP

Catalog No. E0057100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0057100ML 100 mL
E005725ML 25 mL
E0057500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0057100ML Supplier TCI America Supplier No. E0057100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 110-73-6
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00002841
InChI Key MIJDSYMOBYNHOT-UHFFFAOYSA-N
Synonym 2-ethylamino ethanol, ethanol, 2-ethylamino, ethyl ethanolamine, n-ethylethanolamine, 2-hydroxyethyl ethylamine, n-ethylmonoethanolamine, 2-n-monoethylaminoethanol, n-ethylaminoethanol, n-ethyl ethanolamine, 2-ethylamino ethan-1-ol
PubChem CID 8072
IUPAC Name 2-(ethylamino)ethanol
SMILES CCNCCO

Specifications

Melting Point -90°C
Color Yellow
Boiling Point 169°C
Quantity 100 mL
UN Number 2922
Formula Weight 89.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Ethylamino)ethanol
EINECSNumber (2)-0295
RTECSNumber KK9100000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.