Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylbutyl acetate, 98%, Thermo Scientific™

Catalog No. AC308141000
Click to view available options
Quantity:
100 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-87-5
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
InChI Key HQLKZWRSOHTERR-UHFFFAOYSA-N
Synonym acetic acid, 2-ethylbutyl ester, 2-ethyl butyl acetate, beta-ethylbutyl acetate, acetic acid 2-ethylbutyl ester, 1-butanol, 2-ethyl-, acetate, fema no. 2425, 2-ethylbutylacetat, 2-ethylbutyl=acetate, acmc-2097ot, 2-ethylbutyl acetate
PubChem CID 61448
IUPAC Name 2-ethylbutyl acetate
SMILES CCC(CC)COC(=O)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.876g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Refractive Index 1.4090 to 1.4120 (20°C, 589nm)
Linear Formula CH3COCHCH(CH5)
Quantity 100 g
Formula Weight 144.21
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2-Ethylbutyl acetate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.