Learn More
2-Ethylhexane-1,3-diol, 97%
Description
2-Ethylhexane-1,3-diol is used in organic synthesis and can be used to produce polyester resin, polyester plasticizers, polyurethane resin. It is also effective vermifuge to mosquitoes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Ethylhexane-1,3-diol is used in organic synthesis and can be used to produce polyester resin, polyester plasticizers, polyurethane resin. It is also effective vermifuge to mosquitoes.
Solubility
Soluble in water (42g/L at 20°C). Soluble in alcohol and ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is hygroscopic in nature.

Specifications
Specifications
Melting Point | -40°C |
Boiling Point | 241°C to 249°C |
Odor | Odorless |
Linear Formula | CH3CH2CH2CHOHCH(CH2CH3)CH2OH |
Quantity | 2500 mL |
Solubility Information | Soluble in water (42g/L at 20°C). Soluble in alcohol and ether. |
Formula Weight | 146.23 |
Percent Purity | 97% |
Chemical Name or Material | 2-Ethylhexane-1,3-diol |
Safety and Handling
Hazard Category | H302-H312-H317-H318 |
Hazard Statement | GHS H Statement H318-H302-H312 Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. |
Precautionary Statement | P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P333+P313-P363-P501c |
EINECSNumber | 202-377-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.