Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thallic acetate, 98%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Specifications
Specifications
| Melting Point | 182°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 25 g |
| Assay Percent Range | 98% |
| Linear Formula | (CH3CO2)3Tl |
| Beilstein | 02, 96 |
| Fieser | 01,1150; 02,406; 03,286; 04,492; 05,655; 07,360 |
| Merck Index | 15, 9406 |
| Solubility Information | Solubility in water: decomposes |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Fatal if swallowed. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 219-913-2 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction