Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Ethylhexyl)magnesium bromide, 1M solution in diethyl ether, AcroSeal™

Catalog No. AC435951000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435951000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC435951000 Supplier Thermo Scientific Chemicals Supplier No. 435951000

CAS: 90224-21-8 | C8H17BrMg | 217.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90224-21-8 , 60-29-7
Molecular Formula C8H17BrMg
Molecular Weight (g/mol) 217.43
MDL Number MFCD02260220
InChI Key NDCJRLBSQQDNOO-UHFFFAOYNA-M
Synonym 2-ethylhexylmagnesium bromide, 2-ethylhexyl magnesium bromide, 2-ethylhexylmagensium bromide, 2-ethylhexylmagnesiumbromide, 2-ethylhexyl-magnesiumbromide, 2-ethylhexylmagnesium-bromide, 2-ethylhexyl-magnesium bromide, ndcjrlbsqqdnoo-uhfffaoysa-m, grignard reagent
PubChem CID 14915511
IUPAC Name bromo(2-ethylhexyl)magnesium
SMILES CCCCC(CC)C[Mg]Br

Specifications

CAS Min % 23.94, 72.29
CAS Max % 27.71, 76.07
Color Colorless to Gray
Density 0.8630g/mL
Flash Point −25°C
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Specific Gravity 0.863
Solubility Information Solubility in water: reacts.
Formula Weight 217.43
Physical Form Solution
Chemical Name or Material (2-Ethylhexyl)magnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Extremely flammable liquid and vapor.
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Reacts violently with water.
May form explosive peroxides.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.