Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylhexyl methacrylate, 99%, stabilized, ACROS Organics™

Click to view available options
Quantity:
1L
5L
Packaging:
Glass bottle
Plastic drum
688-84-6
C12H22O2
198.31
MFCD00009494
WDQMWEYDKDCEHT-UHFFFAOYSA-N
2-ethylhexyl methacrylate,2-ethyl-1-hexyl methacrylate,2-ethylhexyl methacryate,2-propenoic acid, 2-methyl-, 2-ethylhexyl ester,methacrylic acid 2-ethylhexyl ester,methacrylic acid, 2-ethylhexyl ester,methacrylate, 2-ethylisohexy,2-ethylhexylmethacrylate,dsstox_cid_7293,dsstox_rid_78391
12713
2-ethylhexyl 2-methylprop-2-enoate
CCCCC(CC)COC(=O)C(=C)C
688-84-6
C12H22O2
198.31
MFCD00009494
WDQMWEYDKDCEHT-UHFFFAOYSA-N
2-ethylhexyl methacrylate,2-ethyl-1-hexyl methacrylate,2-ethylhexyl methacryate,2-propenoic acid, 2-methyl-, 2-ethylhexyl ester,methacrylic acid 2-ethylhexyl ester,methacrylic acid, 2-ethylhexyl ester,methacrylate, 2-ethylisohexy,2-ethylhexylmethacrylate,dsstox_cid_7293,dsstox_rid_78391
12713
2-ethylhexyl 2-methylprop-2-enoate
CCCCC(CC)COC(=O)C(=C)C
Boiling Point 120°C (18.0mmHg)
Packaging Glass bottle
Melting Point -50.0°C
Infrared Spectrum Authentic
Flash Point 92°C
Density 0.8840g/mL
Quantity 1L
Specific Gravity 0.884
Stabilizer 40 to 60ppm MEHQ
Color Undesignated
Refractive Index 1.4370 to 1.4390
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula H2C=C(CH3)CO2CH2CH(C2H5)(CH2)3CH3
Solubility Information Solubility in water: 0.0031g/L
Formula Weight 198.31
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-Ethylhexyl methacrylate
Show More Show Less

GHS H StatementMay cause an allergic skin reaction.May cause respiratory irritation.Causes skin irritation.Causes serious eye irritation.Harmful to aquatic life with long lasting effects.

GHS P StatementIF ON SKIN: Wash with plenty of soap and water.If skin irritation or rash occurs: Get medical advice/attention.If eye irritation persists: Get medical advice/attention.IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.Call a POISON CENTER or doctor/physician if you feel unwell.

;GHS P StatementIF ON SKIN: Wash with plenty of soap and water.If skin irritation or rash occurs: Get medical advice/attention.If eye irritation persists: Get medical advice/attention.IF INHALED: Remove to fresh air and keep at rest in a po