Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylhexylamine 98.0+%, TCI America™
SDP

Catalog No. E0126100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0126100ML 100 mL
E012625ML 25 mL
E0126500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0126100ML Supplier TCI America Supplier No. E0126100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 104-75-6
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00008148
InChI Key LTHNHFOGQMKPOV-UHFFFAOYSA-N
Synonym 2-ethylhexylamine, 1-hexanamine, 2-ethyl, 2-ethyl-1-hexylamine, 2-ethyl hexylamine, 2-ethylhexanamine, 1-amino-2-ethylhexane, hexylamine, 2-ethyl, beta-ethylhexylamine, 1-amino-2-ethylhexan, isooctylame
PubChem CID 7719
IUPAC Name 2-ethylhexan-1-amine
SMILES CCCCC(CC)CN

Specifications

Color Colorless
Boiling Point 168°C
Quantity 100 mL
UN Number 2276
Formula Weight 129.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexylamine
EINECSNumber (2)-0133
RTECSNumber MQ5250000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.