Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylphenyl Acetate 97.0+%, TCI America™
SDP

Catalog No. A154925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A154925G 25 g
A15495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A154925G Supplier TCI America Supplier No. A154925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3056-59-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD02093429
InChI Key VERVERDPMMBDSS-UHFFFAOYSA-N
Synonym Acetic Acid 2-Ethylphenyl Ester
PubChem CID 9898919
IUPAC Name (2-ethylphenyl) acetate
SMILES CCC1=CC=CC=C1OC(=O)C

Specifications

Color Yellow
Quantity 25 g
Formula Weight 164.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylphenyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.