Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-3-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. F098225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F098225G 25 g
F09825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F098225G Supplier TCI America Supplier No. F098225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1480-87-1
Molecular Formula C5H3FN2O2
Molecular Weight (g/mol) 142.09
MDL Number MFCD03095068
InChI Key QDKIYDGHCFZBGC-UHFFFAOYSA-N
Synonym 2-fluoro-3-nitro-pyridine, pyridine, 2-fluoro-3-nitro, pubchem1115, acmc-1bwlp, 2-fluor-3-nitropyridine, 2-fluoro-3-nitropyridine, ksc174g2n
PubChem CID 2762817
IUPAC Name 2-fluoro-3-nitropyridine
SMILES [O-][N+](=O)C1=C(F)N=CC=C1

Specifications

Melting Point 18°C
Boiling Point 110°C
Quantity 25 g
Formula Weight 142.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-3-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.