Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-hydroxybenzoic acid, 98%, Thermo Scientific™

Catalog No. p-7077686
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 65145-13-3
Molecular Formula C7H5FO3
Molecular Weight (g/mol) 156.11
MDL Number MFCD01310985
InChI Key NXWTWYULZRDBSA-UHFFFAOYSA-N
Synonym 2-fluoro-4-hydroxy-benzoic acid, benzoic acid, 2-fluoro-4-hydroxy, pubchem2608, acmc-1b9jp, 4-hydroxyfluorobenzoic acid, frinton fr-2445, ksc352q5p, 2-fluoro-4-hydroxybenzoicacid, 4-hydroxy-2-fluorobenzoic acid, 2-fluoro4-hydroxy-benzoic acid
PubChem CID 2783398
IUPAC Name 2-fluoro-4-hydroxybenzoic acid
SMILES OC(=O)C1=C(F)C=C(O)C=C1

Specifications

Melting Point 187.0°C to 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Formula Weight 156.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-hydroxybenzoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.